Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFAF1 Antibody (SR1548), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | NDUFAF1 |
---|---|
Clone | SR1548 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Description
NDUFAF1 Monoclonal antibody specifically detects NDUFAF1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, ImmunoprecipitationSpecifications
NDUFAF1 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CGI-65, CIA30CGI65, complex I intermediate-associated protein 30, mitochondrial, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 1, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 1 | |
A synthesized peptide derived from human NDUFAF1 (Uniprot #: Q9Y375) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1548 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Endocrinology, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
51103 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title