Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFAF1 Recombinant Rabbit Monoclonal Antibody (4K8Y8)
GREENER_CHOICE

Catalog No. PIMA538101
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538101 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538101 Supplier Invitrogen™ Supplier No. MA538101
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: U2OS, 293T, HepG2 Immunogen sequence: MALVHKLLRG TYFLRKFSKP TSALYPFLGI RFAEYSSSLQ KPVASPGKAS SQRKTEGDLQ GDHQKEVALD ITSSEEKPDV SFDKAIRDEA IYHFRLLKDE IVDHWRGPEG HPLHEVLLEQ AKVVWQFRGK EDLDKWTVTS DKTIGGRSEV.

The NDUFAF1 gene encodes the human homolog of a Neurospora crassa Cia30 protein involved in the assembly of complex I.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFAF1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 4K8Y8
Concentration 1.76 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NDUFAF1
Gene Accession No. Q9Y375
Gene Alias 2410001M24Rik; CGI65; CGI-65; CIA30; complex I intermediate-associated protein 30, mitochondrial; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 1; NADH dehydrogenase (ubiquinone) 1 assembly factor 1; NADH dehydrogenase (ubiquinone) complex I, assembly factor 1; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 1; NADH:ubiquinone oxidoreductase complex assembly factor 1; NADH-ubiquinone oxidoreductase 1 alpha subcomplex, assembly factor 1; Ndufaf1
Gene Symbols NDUFAF1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-150 of human NDUFAF1 (Q9Y375).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51103
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.