Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NDUFAF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598667
Description
NDUFAF2 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Mimitin, a small mitochondrial protein, whose transcription is directly stimulated by c-Myc, is highly expressed in 80% of esophageal squamous cell carcinomas (ESCC). Suppression of Mimitin expression by RNA interference had no effect in cancerous cell lines such as human cervical carcinoma or hepatocarcinoma cell lines, but caused a decrease in cell proliferation in human glioblastoma, embryonic lung fibroblastic cells, and ESCC, suggesting Mimitin may play a special role in these types of cells. Mimitin expression is also regulated by MAPK kinases and IL-1, but not through the NF-kappa-B-related pathway. It will interact with the microtubular protein MAP1S and can affect the activities of caspase-3 and -7 in cells stimulated to develop apoptosis. Other experiments suggest that Mimitin also acts as a molecular chaperone for the assembly of the mitochondrial complex I.
Specifications
NDUFAF2 | |
Polyclonal | |
Unconjugated | |
Ndufaf2 | |
1810058I14Rik; B17.2L; B17.2-like; B172L; B172-like; C86051; mimitin; mimitin, mitochondrial; MMTN; Myc-induced mitochondria protein; Myc-induced mitochondrial protein; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 2; NADH dehydrogenase (ubiquinone) complex I, assembly factor 2; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 2; NADH dehydrogenase 1 alpha subcomplex assembly factor 2; NADH:ubiquinone oxidoreductase complex assembly factor 2; NDUFA12L; NDUFA12-like protein; Ndufaf2; RGD1560158 | |
Rabbit | |
Protein G | |
RUO | |
91942 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
2.78 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q8N183 | |
Ndufaf2 | |
Recombinant Human NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 2 protein (117-169AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction