Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFAF3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557250
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557250 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557250 Supplier Invitrogen™ Supplier No. PA557250
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLEPRIEIVV VGTGDRTERL QSQVLQAMRQ RGIAVEVQDT PNACATFNFL CHEGRVTGAA LIPPPGGTSL TSLGQAAQ Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 85%.

This gene encodes a mitochondrial complex I assembly protein that interacts with complex I subunits. Mutations in this gene cause mitochondrial complex I deficiency, a fatal neonatal disorder of the oxidative phosphorylation system. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFAF3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NDUFAF3
Gene Accession No. O08776, Q9BU61
Gene Alias 2P1; 4733401H18Rik; C3orf60; E3-3; fc05g04; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 3; NADH dehydrogenase (ubiquinone) complex I, assembly factor 3; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 3; NADH:ubiquinone oxidoreductase complex assembly factor; NADH:ubiquinone oxidoreductase complex assembly factor 3; NDUFAF3; Nuclear protein E3-3; protein 2P1; RGD708545; wu:fc05g04; zgc:153176
Gene Symbols NDUFAF3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NDUFAF3. Recombinant protein control fragment (Product #RP-96452).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25915, 56769
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.