Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFAF4 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980887
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0887 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0887 Supplier Bioss Supplier No. BS10456R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NADH:ubiquinone oxidoreductase (complex I) catalyzes the transfer of electrons from NADH to ubiquinone (coenzyme Q) in the first step of the mitochondrial respiratory chain, resulting in the translocation of protons across the inner mitochondrial membrane. This gene encodes a complex I assembly factor. Mutations in this gene are a cause of mitochondrial complex I deficiency.

Specifications

Antigen NDUFAF4
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NDUFAF4
Gene Accession No. Q9P032
Gene Alias 1110007M04Rik; 3000003G13Rik; AW214064; bA22L21.1; C6orf66; Hormone-regulated proliferation-associated protein of 20 kDa; Hormone-regulated proliferation-associated protein of 20 kDa homolog; hormone-regulated proliferation-associated protein, 20 kDa; Hrpap20; HSPC125; My013; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 4; NADH dehydrogenase (ubiquinone) complex I, assembly factor 4; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 4; NADH:ubiquinone oxidoreductase complex assembly factor 4; Ndufaf4
Gene Symbols NDUFAF4
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NDUFAF4.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29078
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.