Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFAF6 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA561446
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561446 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561446 Supplier Thermo Scientific Supplier No. PA561446

Rabbit Polyclonal Antibody

Immunogen sequence: LAQVKDSVSE KTIGLMRMQF WKKTVEDIYC DNPPHQPVAI ELWKAVKRHN LTKRWLMKIV DEREKNLDDK AYRNIKELEN YA Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 87%.

NDUFAF6 Polyclonal antibody specifically detects NDUFAF6 in Human samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFAF6
Applications Immunohistochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NDUFAF6
Gene Accession No. Q330K2
Gene Alias C8orf38; NADH dehydrogenase (ubiquinone) complex I, assembly factor 6; NADH:ubiquinone oxidoreductase complex assembly factor 6; NDUFAF6; Putative phytoene synthase; UPF0551 protein C8orf38, mitochondrial
Gene Symbols NDUFAF6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NDUFAF6
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 137682
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.