Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NDUFAF7 Monoclonal Antibody (OTI4A5)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA525564
Description
NDUFAF7 Monoclonal Antibody for Western Blot, IHC (P)
The function of this protein remains unknown.
Specifications
NDUFAF7 | |
Monoclonal | |
0.86 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q7L592 | |
NDUFAF7 | |
Full length human recombinant protein of C2orf56 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4A5 | |
Unconjugated | |
NDUFAF7 | |
2410091C18Rik; AL033374; C2orf56; MidA; mitochondrial dysfunction protein A homolog; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex assembly factor 7; NADH dehydrogenase (ubiquinone) complex I, assembly factor 7; NADH dehydrogenase [ubiquinone] complex I, assembly factor 7; NADH:ubiquinone oxidoreductase complex assembly factor 7; NDUFAF7; PRO1853; Protein arginine methyltransferase NDUFAF7, mitochondrial; Protein midA homolog; protein midA homolog, mitochondrial; RGD1311578 | |
Mouse | |
Affinity Chromatography | |
RUO | |
55471 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction