Learn More
Invitrogen™ NDUFAF7 Polyclonal Antibody

Description
Immunogen sequence: ISVHLVEVSQ KLSEIQALTL TKEKVPLERN AGSPVYMKGV TKSGIPISWY RDLHDVPKGY SFYLAHEFFD VLPVHKFQKT PQGWREVFVD IDPQVSD Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 89%.
Specifications
Specifications
| Antigen | NDUFAF7 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | NDUFAF7 |
| Gene Accession No. | Q7L592 |
| Gene Alias | 2410091C18Rik; AL033374; C2orf56; MidA; mitochondrial dysfunction protein A homolog; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex assembly factor 7; NADH dehydrogenase (ubiquinone) complex I, assembly factor 7; NADH dehydrogenase [ubiquinone] complex I, assembly factor 7; NADH:ubiquinone oxidoreductase complex assembly factor 7; NDUFAF7; PRO1853; Protein arginine methyltransferase NDUFAF7, mitochondrial; Protein midA homolog; protein midA homolog, mitochondrial; RGD1311578 |
| Gene Symbols | NDUFAF7 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.