Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116705
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116705 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116705 Supplier Invitrogen™ Supplier No. PA5116705
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: 293T, A375, Mouse brain, Mouse pancreas Immunogen sequence: MICWRHPSAP CGRGEWQVPR SQLPLARVEF PVALGLGVAV GAEAAAIMVN LLQIVRDHWV HVLVPMGFVI GCYLDRKSDE RLTAFRNKSM LFKRELQPSE EVTWK.

NDUFB1 (NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 1), also known as CI-MNLL (complex I-MNLL), CI-SGDH or NADH-ubiquinone oxidoreductase MNLL subunit, is a 58 amino acid single-pass membrane protein that localizes to the matrix side of the mitochondrial membrane. A member of the complex I NDUFB1 subunit family, NDUFB1 is encoded by a gene that maps to human chromosome 14q32.12. Chromosome 14 houses over 700 genes and comprises nearly 3.5% of the human genome. Chromosome 14 encodes the presinilin 1 (PSEN1) gene, which is one of the three key genes associated with the development of Alzheimer's disease (AD). The SERPINA1 gene is also located on chromosome 14 and, when defective, leads to the genetic disorder alpha1-antitrypsin deficiency, which is characterized by severe lung complications and liver dysfunction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFB1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.98 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene NDUFB1
Gene Accession No. O75438, P0DN34
Gene Alias CI-MNLL; CI-SGDH; complex I MNLL subunit; complex I-MNLL; MNLL; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 1; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 1, 7kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 1; NADH:ubiquinone oxidoreductase subunit B1; NADH-ubiquinone oxidoreductase MNLL subunit; Ndufb1
Gene Symbols NDUFB1
Host Species Rabbit
Immunogen Recombinant Protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110324555, 4707
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.