Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFB1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | NDUFB1 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-1247
![]() ![]() |
Proteintech Group Inc
169021AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
NDUFB1 (NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 1), also known as CI-MNLL (complex I-MNLL), CI-SGDH or NADH-ubiquinone oxidoreductase MNLL subunit, is a 58 amino acid single-pass membrane protein that localizes to the matrix side of the mitochondrial membrane. A member of the complex I NDUFB1 subunit family, NDUFB1 is encoded by a gene that maps to human chromosome 14q32.12. Chromosome 14 houses over 700 genes and comprises nearly 3.5% of the human genome. Chromosome 14 encodes the presinilin 1 (PSEN1) gene, which is one of the three key genes associated with the development of Alzheimer′s disease (AD). The SERPINA1 gene is also located on chromosome 14 and, when defective, leads to the genetic disorder alpha1-antitrypsin deficiency, which is characterized by severe lung complications and liver dysfunction.Specifications
NDUFB1 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
O75438 | |
4707 | |
NDUFB1 Fusion Protein Ag10443 | |
Primary | |
-20°C | |
NDUFB1 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CI MNLL, CI SGDH, Complex I MNLL, MNLL, NDUFB1 | |
NDUFB1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title