Learn More
Invitrogen™ NDUFB10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596266
Description
Immunogen sequence: MPDSWDKDVY PEPPRRTPVQ PNPIVYMMKA FDLIVDRPVT LVREFIERQH AKNRYYYYHR QYRRVPDITE CKEEDIMCMY EAEMQWKRDY KVDQEIINIM QDRLKACQQR EGQNYQQNCI KEVEQFTQVA KAYQDRYQDL GAYSSARKCL AKQRQRMLQE RKAAKEAAAA TS; Positive Samples: HepG2, HeLa, MCF-7, Jurkat, Mouse brain, Mouse kidney, Mouse heart; Cellular Location: Matrix side, Mitochondrion inner membrane, Peripheral membrane protein.
NDUFB10 is an accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. Complex I is composed of 45 different subunits.
Specifications
NDUFB10 | |
Polyclonal | |
Unconjugated | |
NDUFB10 | |
0610011B04Rik; 22kDa; CI PDSW; CI-PDSW; Complex I PDSW; complex I PDSW subunit; complex I-PDSW; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 10; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 10, 22kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 10; NADH ubiquinone oxidoreductase PDSW subunit (RH 16p13.3); NADH:ubiquinone oxidoreductase subunit B10; NADH-ubiquinone oxidoreductase PDSW subunit; NDUFB 10; NDUFB10; PDSW | |
Rabbit | |
Affinity Chromatography | |
RUO | |
4716, 681418, 68342 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
1.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O96000, Q9DCS9 | |
NDUFB10 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-172 of human NDUFB10 (NP_0045391). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.