Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFB11 Antibody (OTI9C5), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272916H
Description
NDUFB11 Monoclonal specifically detects NDUFB11 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
NDUFB11 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
NDUFB11 | |
Recombinant protein fragment corresponding to amino acids 30-163 of human NDUFB11 (NP_061929) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI9C5 | |
Western Blot, Immunohistochemistry | |
CI-ESSS, complex I NP17.3 subunit, Complex I-ESSS, ESSS, FLJ20494, MGC111182, NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 11, 17.3kDa, NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 11, mitochondrial, NADH-ubiquinone oxidoreductase ESSS subunit, Neuronal protein 17.3, Np15, Np17.3, P17.3 | |
Mouse | |
Affinity Purified | |
Stem Cell Markers | |
54539.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction