Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFB11 Monoclonal Antibody (OTI8C5), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA807693

Description
NDUFB11 is an accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) that is believed not to be involved in catalysis. NDUFB11 functions in the transfer of electrons from NADH to the respiratory chain and is located in the inner mitochondrial membrane. The immediate electron acceptor for NDUFB11 is believed to be ubiquinone. Further, NDUFB11 is believed to have NADH dehydrogenase activity, and oxidoreductase activity. Diseases associated with NDUFB11 protein dysfunction include linear skin defects with multiple congenital anomalies and mitochondrial complex I deficiency.Specifications
NDUFB11 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9NX14 | |
NDUFB11 | |
Human recombit protein fragment corresponding to amino acids 30-163 of human NDUFB11 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI8C5 | |
Unconjugated | |
NDUFB11 | |
CI-ESSS, ESSS, NP17.3, Np15, P17.3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
54539 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction