Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFB11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556449
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556449 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556449 Supplier Invitrogen™ Supplier No. PA556449
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PDYRMKEWSR REAERLVKYR EANGLPIMES NCFDPSKIQL PEDE Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.

NDUFB11 is an accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) that is believed not to be involved in catalysis. NDUFB11 functions in the transfer of electrons from NADH to the respiratory chain and is located in the inner mitochondrial membrane. The immediate electron acceptor for NDUFB11 is believed to be ubiquinone. Further, NDUFB11 is believed to have NADH dehydrogenase activity, and oxidoreductase activity. Diseases associated with NDUFB11 protein dysfunction include linear skin defects with multiple congenital anomalies and mitochondrial complex I deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFB11
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NDUFB11
Gene Accession No. Q9NX14
Gene Alias CI-ESSS; complex I NP17.3 subunit; complex I-ESSS; D5Bwg0566e; D5Bwg0577e; ESSS; LSDMCA3; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 11; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 11, 17.3kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 11, mitochondrial; NADH:ubiquinone oxidoreductase subunit B11; NADH-ubiquinone oxidoreductase ESSS subunit; NDUFB11; Neuronal protein 15.6; neuronal protein 17.3; Np15; NP15.6; Np17.3; nuclear protein 15.6; p15.6; P17.3; RGD1563698; UNQ111/PRO1064
Gene Symbols NDUFB11
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NDUFB11. Recombinant protein control fragment (Product #RP-95188).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54539
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.