Learn More
Invitrogen™ NDUFB11 Polyclonal Antibody

Description
Immunogen sequence: MAAGLFGLSA RRLLAAAATR GLPAARVRWE SSFSRTVVAP SAVAGKRPPE PTTPWQEDPE PEDENLYEKN PDSHGYDKDP VLDVWNMRLV FFFGVSIILV LGSTFVAYLP DYRCTGCPRA WDGMKEWSRR EAERLVKYRE ANGLPIMESN CFDPSKIQLP EDE; Positive Samples: HepG2, U-251MG, K-562, HeLa, Mouse liver, Mouse heart, Rat brain, Rat liver.
Specifications
Specifications
| Antigen | NDUFB11 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.11 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 |
| Gene | NDUFB11 |
| Gene Accession No. | O09111, Q9NX14 |
| Gene Alias | CI-ESSS; complex I NP17.3 subunit; complex I-ESSS; D5Bwg0566e; D5Bwg0577e; ESSS; LSDMCA3; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 11; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 11, 17.3kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 11, mitochondrial; NADH:ubiquinone oxidoreductase subunit B11; NADH-ubiquinone oxidoreductase ESSS subunit; NDUFB11; Neuronal protein 15.6; neuronal protein 17.3; Np15; NP15.6; Np17.3; nuclear protein 15.6; p15.6; P17.3; RGD1563698; UNQ111/PRO1064 |
| Gene Symbols | NDUFB11 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.