Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100671
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100671 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100671 Supplier Invitrogen™ Supplier No. PA5100671
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NDUFB2.

The protein encoded by this gene belongs to the peptidase family S14 and hydrolyzes proteins into small peptides in the presence of ATP and magnesium. The protein is transported into mitochondrial matrix and is associated with the inner mitochondrial membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFB2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NDUFB2
Gene Accession No. O95178, Q9CPU2
Gene Alias 1810011O01Rik; 8kDa; AGGG; AI325567; CI-AGGG; complex I AGGG subunit; complex I-AGGG; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 2; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 2, 8kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 2, mitochondrial; NADH:ubiquinone oxidoreductase subunit B2; NADH-ubiquinone oxidoreductase AGGG subunit; NDUFB2
Gene Symbols NDUFB2
Host Species Rabbit
Immunogen A synthesized peptide derived from human NDUFB2(Accession O95178), corresponding to amino acid residues A3-S53.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4708, 68198
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.