Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562394
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562394 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562394 Supplier Invitrogen™ Supplier No. PA562394
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QVFQSEFFSG LMWFWILWRF WHDSEEVLGH FPYPDPSQWT DEELGIPPDD ED Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 79%.

The protein encoded by this gene belongs to the peptidase family S14 and hydrolyzes proteins into small peptides in the presence of ATP and magnesium. The protein is transported into mitochondrial matrix and is associated with the inner mitochondrial membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFB2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NDUFB2
Gene Accession No. O95178
Gene Alias 1810011O01Rik; 8kDa; AGGG; AI325567; CI-AGGG; complex I AGGG subunit; complex I-AGGG; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 2; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 2, 8kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 2, mitochondrial; NADH:ubiquinone oxidoreductase subunit B2; NADH-ubiquinone oxidoreductase AGGG subunit; NDUFB2
Gene Symbols NDUFB2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NDUFB2. Recombinant protein control fragment (Product #RP-100093).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4708
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.