Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529258
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529258 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529258 Supplier Invitrogen™ Supplier No. PA529258
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: mouse Lung. Predicted reactivity: Mouse (81%), Chimpanzee (100%), Bovine (82%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The multisubunit NADH:ubiquinone oxidoreductase (complex I) is the first enzyme complex in the electron transport chain of mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFB3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene NDUFB3
Gene Accession No. O43676, Q9CQZ6
Gene Alias 2700033I16Rik; AI415450; B12; CI-B12; complex I-B12; NADH dehydrogenase (ubiquinone) 1 beta subcomplex 3; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 3, 12kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 3; NADH:ubiquinone oxidoreductase subunit B3; NADH-ubiquinone oxidoreductase B12 subunit; Ndufb3
Gene Symbols NDUFB3
Host Species Rabbit
Immunogen Full length human NDUFB3 Recombinant protein.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4709, 66495
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.