Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556986
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556986 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556986 Supplier Invitrogen™ Supplier No. PA556986
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAHEHGHEHG HHKMELPDYR QWKIEGTPLE TIQKKLAAKG LRDPWGRNEA WRYMGGFAKS VSF Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 80%.

The multisubunit NADH:ubiquinone oxidoreductase (complex I) is the first enzyme complex in the electron transport chain of mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFB3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NDUFB3
Gene Accession No. O43676
Gene Alias 2700033I16Rik; AI415450; B12; CI-B12; complex I-B12; NADH dehydrogenase (ubiquinone) 1 beta subcomplex 3; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 3, 12kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 3; NADH:ubiquinone oxidoreductase subunit B3; NADH-ubiquinone oxidoreductase B12 subunit; Ndufb3
Gene Symbols NDUFB3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NDUFB3. Recombinant protein control fragment (Product #RP-96049).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4709
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.