Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFB3 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549506
Description
The multisubunit NADH:ubiquinone oxidoreductase (complex I) is the first enzyme complex in the electron transport chain of mitochondria. See NDUFA2 (MIM 602137).
Specifications
| NDUFB3 | |
| Polyclonal | |
| PBS with 0.09% sodium azide | |
| O43676, Q9CQZ6 | |
| NDUFB3 | |
| KLH conjugated synthetic peptide between 13-45 amino acids from human NDUFB3. | |
| 200 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| NDUFB3 | |
| NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 3, Complex I-B12, CI-B12, NADH-ubiquinone oxidoreductase B12 subunit, NDUFB3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4709, 66495 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction