Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFB4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545913
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545913 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545913 Supplier Invitrogen™ Supplier No. PA545913
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ARTINVYPNF RPTPKNSLMG ALCGFGPLIF IYYIIKTERD RKEKLIQEGK Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 79%; Horse: 86%; Human: 100%; Mouse: 86%; Pig: 86%; Rat: 86%.

The protein encoded by this gene belongs to the complex I NDUFB4 subunit family. Mammalian complex I is composed of 45 different subunits. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFB4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NDUFB4
Gene Accession No. O95168
Gene Alias 0610006N12Rik; 1300010H20Rik; B15; B15 subunit of the NADH: ubiquinone oxidoreductase complex; CI-B15; complex I B15 subunit; complex I-B15; MGC5105; NADH dehydrogenase (ubiquinone) 1 beta subcomplex 4; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 4, 15kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 4; NADH:ubiquinone oxidoreductase B15 subunit; NADH:ubiquinone oxidoreductase subunit B4; NADH-ubiquinone oxidoreductase B15 subunit; Ndufb4; similar to B15 subunit of the NADH: ubiquinone oxidoreductase complex
Gene Symbols NDUFB4
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human NDUFB4 (aa 71-120).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4710
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.