Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFB4 Recombinant Rabbit Monoclonal Antibody (JE65-30)
GREENER_CHOICE

Catalog No. PIMA544696
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544696 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA544696 Supplier Invitrogen™ Supplier No. MA544696
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

NDUFB4 Recombinant Monoclonal Antibody for IHC (P), Western Blot

The protein encoded by this gene belongs to the complex I NDUFB4 subunit family. Mammalian complex I is composed of 45 different subunits. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFB4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE65-30
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene NDUFB4
Gene Accession No. O95168, Q9CQC7
Gene Alias 0610006N12Rik; 1300010H20Rik; B15; B15 subunit of the NADH: ubiquinone oxidoreductase complex; CI-B15; complex I B15 subunit; complex I-B15; MGC5105; NADH dehydrogenase (ubiquinone) 1 beta subcomplex 4; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 4, 15kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 4; NADH:ubiquinone oxidoreductase B15 subunit; NADH:ubiquinone oxidoreductase subunit B4; NADH-ubiquinone oxidoreductase B15 subunit; Ndufb4; similar to B15 subunit of the NADH: ubiquinone oxidoreductase complex
Gene Symbols NDUFB4
Host Species Rabbit
Immunogen Recombinant protein within human NDUFB4 aa 1-129/129.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 288088, 4710, 68194
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.