Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFB5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552172
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552172 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552172 Supplier Invitrogen™ Supplier No. PA552172
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EGYVPEHWEY YKHPISRWIA RNFYDSPEKI YERTMAVLQI EAEKAELRVK ELEVRKLMHV RGDGPWYYYE TIDKELIDHS PKATPDN Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 77%.

NDUFB5 is a subunit of the multisubunit NADH:ubiquinone oxidoreductase (complex I). Mammalian complex I is composed of 45 different subunits. It locates at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. The protein encoded by this gene is a subunit of the multisubunit NADH:ubiquinone oxidoreductase (complex I). Mammalian complex I is composed of 45 different subunits. It locates at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFB5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NDUFB5
Gene Accession No. O43674
Gene Alias 0610007D05Rik; 16 kDa precursor; AU015782; CISGDH; CI-SGDH; complex I SGDH subunit; complex I-SGDH; DKFZp686N02262; FLJ30597; hm:zeh0024; hypothetical protein LOC557853; MGC111204; MGC12314; NADH dehydrogenase; NADH dehydrogenase (ubiquinone) 1 beta subcomplex 5; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 5; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 5, 16kDa; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 5, 16kDa, precursor; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 5, mitochondrial; NADH dehydrogenase 1 beta subcomplex 5; NADH:ubiquinone oxidoreductase subunit B5; NADH-ubiquinone oxidoreductase SGDH subunit; Ndufb5; SGDH; zgc:123331
Gene Symbols NDUFB5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NDUFB5. Recombinant protein control fragment (Product #RP-102248).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4711
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.