Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFB5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598000
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598000 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598000 Supplier Invitrogen™ Supplier No. PA598000
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NDUFB5 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

NDUFB5 is a subunit of the multisubunit NADH:ubiquinone oxidoreductase (complex I). Mammalian complex I is composed of 45 different subunits. It locates at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. The protein encoded by this gene is a subunit of the multisubunit NADH:ubiquinone oxidoreductase (complex I). Mammalian complex I is composed of 45 different subunits. It locates at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFB5
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NDUFB5
Gene Accession No. O43674, Q9CQH3
Gene Alias 0610007D05Rik; 16 kDa precursor; AU015782; CISGDH; CI-SGDH; complex I SGDH subunit; complex I-SGDH; DKFZp686N02262; FLJ30597; hm:zeh0024; hypothetical protein LOC557853; MGC111204; MGC12314; NADH dehydrogenase; NADH dehydrogenase (ubiquinone) 1 beta subcomplex 5; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 5; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 5, 16kDa; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 5, 16kDa, precursor; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 5, mitochondrial; NADH dehydrogenase 1 beta subcomplex 5; NADH:ubiquinone oxidoreductase subunit B5; NADH-ubiquinone oxidoreductase SGDH subunit; Ndufb5; SGDH; zgc:123331
Gene Symbols NDUFB5
Host Species Rabbit
Immunogen Recombinant Human NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 5, mitochondrial protein (100-189AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4711, 66046
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.