Learn More
Invitrogen™ NDUFB6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier:  Invitrogen™ PA551308
Catalog No. PA551308
Only null left
Add to Cart
Description
Description
The antibody detects endogenous levels of total NDUFB6 protein.
Methylation of cytosine at CpG dinucleotides is a common feature of many higher eukaryotic genomes. Increasing evidence indicates that recruitement of histone deacetylase complexes by methyl-CpG-binding proteins (MeCP) is a major mechanism of methylated DNA silencing. P66, a zinc finger-containing protein is one of the components of the MeCP1 complex. P66 and p68, the two components of MeCP1 are different forms of the same zinc finger-containing protein and are conserved from C.elegans to humans. Homologs of p66 from different organisms revealed two highly conserved regions, CR1 and CR2. While CR1 is involved in the association of p66 with other MeCP1 components, CR2 plays an important role in targeting p66 and MBD3 to specific loci.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NDUFB6
Polyclonal
Unconjugated
NDUFB6
B17; CI; CI-B17; complex I, mitochondrial respiratory chain, B17 subunit; complex I-B17; Gm137; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 6; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 6, 17kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 6; NADH:ubiquinone oxidoreductase subunit B6; NADH-ubiquinone oxidoreductase B17 subunit; NADH-ubiquinone oxidoreductase beta subunit, 6; NDUFB6
Rabbit
Antigen affinity chromatography
RUO
4712
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
O95139
NDUFB6
Full length fusion protein of human NDUFB6.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.