Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFB7 Rabbit anti-Human, HRP, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299124H
Description
NDUFB7 Polyclonal specifically detects NDUFB7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
NDUFB7 | |
Polyclonal | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HRP | |
B18, Cell adhesion protein SQM1, CI-B18NADH-ubiquinone oxidoreductase B18 subunit, complex I B18 subunit, Complex I-B18, MGC2480, NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 7 (18kD, B18), NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 7, 18kDa, NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 7 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human NDUFB7. | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
4713 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction