Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFB8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103949
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103949 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103949 Supplier Invitrogen™ Supplier No. PA5103949
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total NDUFB8.

Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFB8
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NDUFB8
Gene Accession No. O95169, Q9D6J5
Gene Alias 19 kDa; 2900010I05Rik; AI987932; ASHI; CI-ASHI; complex I ASHI subunit; complex I-ASHI; NADH dehydrogenase; NADH dehydrogenase (ubiquinone) 1 beta subcomplex 8; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 8, 19kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 8, mitochondrial; NADH dehydrogenase 1 beta subcomplex 8; NADH:ubiquinone oxidoreductase ASHI subunit; NADH:ubiquinone oxidoreductase subunit B8; NADH-ubiquinone oxidoreductase ASHI subunit; NDUFB8
Gene Symbols NDUFB8
Host Species Rabbit
Immunogen A synthesized peptide derived from human NDUFB8(Accession O95169), corresponding to amino acid residues G39-W89.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 293991, 4714, 67264
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.