Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFB9 Antibody (OTI13H11), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272919AF488
Description
NDUFB9 Monoclonal specifically detects NDUFB9 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
| NDUFB9 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NDUFB9 | |
| Recombinant protein fragment corresponding to amino acids 3-179 of human NDUFB9 (NP_0049906) produced in E.coli. | |
| 0.1 mL | |
| 4715.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI13H11 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| B22, CI-B22, complex I B22 subunit, Complex I-B22, FLJ22885, LYR motif-containing protein 3, LYRM3DKFZp566O173, NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9, 22kDa, NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 9, NADH-ubiquinone oxidoreductase B22 subunit, UQOR22NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9 (22kD, B22) | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction