Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFB9 Antibody (OTI8B7), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272918APCCY7
Description
NDUFB9 Monoclonal specifically detects NDUFB9 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Flow Cytometry.Specifications
NDUFB9 | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
NDUFB9 | |
Recombinant protein fragment corresponding to amino acids 3-179 of human NDUFB9 (NP_001893) produced in E.coli. | |
0.1 mL | |
4715.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI8B7 | |
Flow Cytometry | |
B22, CI-B22, complex I B22 subunit, Complex I-B22, FLJ22885, LYR motif-containing protein 3, LYRM3DKFZp566O173, NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9, 22kDa, NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 9, NADH-ubiquinone oxidoreductase B22 subunit, UQOR22NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9 (22kD, B22) | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Monkey, Primate | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction