Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFB9 Monoclonal Antibody (OTI8B7), TrueMAB™, OriGene
SDP

Catalog No. 502541457
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1457 30 μL
50-167-3613 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1457 Supplier OriGene Supplier No. TA502567S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NDUFB9 Monoclonal antibody specifically detects NDUFB9 in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen NDUFB9
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI8B7
Concentration 0.76 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MEF2C
Gene Accession No. Q06413
Gene Alias 1190008J14Rik; B22; CI-B22; complex I B22 subunit; complex I-B22; EGK_19260; LYR motif-containing protein 3; LYRM3; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9, 22kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 9; NADH:ubiquinone oxidoreductase subunit B9; NADH-ubiquinone oxidoreductase B22 subunit; NDUFB9; UQOR22
Host Species Mouse
Immunogen Full length human recombinant protein of human MEF2C produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4208
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.