Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFC1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP15476020UL
Description
NDUFC1 Polyclonal specifically detects NDUFC1 in Human samples. It is validated for Western Blot.Specifications
NDUFC1 | |
Polyclonal | |
Western Blot 1:100-1:2000 | |
O43677 | |
NDUFC1 | |
Synthetic peptides corresponding to NDUFC1(NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 1, 6kDa) The peptide sequence was selected from the middle region of NDUFC1. Peptide sequence RSKFYVREPPNAKPDWLKVGFTLGTTVFLWIYLIKQHNEDILEYKRR | |
Affinity Purified | |
RUO | |
4717 | |
Human | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
complex I KFYI subunit, KFYI, MGC117464, MGC126847, MGC138266, NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 1, 6kDa, NADH dehydrogenase [ubiquinone] 1 subunit C1, mitochondrial, subcomplex unknown, 1 (6kD, KFYI) | |
Rabbit | |
9 kDa | |
20 μL | |
Primary | |
This product is specific to Subunit or Isoform: C1, mitochondrial. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction