Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560747
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560747 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560747 Supplier Invitrogen™ Supplier No. PA560747
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSGPSVRSKF YVREPPNAKP DWLKVG Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 77%.

Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFC1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NDUFC1
Gene Accession No. O43677
Gene Alias 2310016K22Rik; CI-KFYI; complex I KFYI subunit; complex I-KFYI; KFYI; NADH dehydrogenase; NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 1; NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 1 (6kD, KFYI); NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 1, 6kDa; NADH dehydrogenase [ubiquinone] 1 subunit C1, mitochondrial; NADH:ubiquinone oxidoreductase subunit C1; NADH-ubiquinone oxidoreductase KFYI subunit; Ndufc1
Gene Symbols NDUFC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NDUFC1. Recombinant protein control fragment (Product #RP-96648).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4717
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.