Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568240
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568240 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568240 Supplier Invitrogen™ Supplier No. PA568240
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Mouse samples.

Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFC1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.87% NaCl, 30% glycerol and 0.01% sodium azide; pH 7.3
Gene NDUFC1
Gene Accession No. O43677, Q9CQY9
Gene Alias 2310016K22Rik; CI-KFYI; complex I KFYI subunit; complex I-KFYI; KFYI; NADH dehydrogenase; NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 1; NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 1 (6kD, KFYI); NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 1, 6kDa; NADH dehydrogenase [ubiquinone] 1 subunit C1, mitochondrial; NADH:ubiquinone oxidoreductase subunit C1; NADH-ubiquinone oxidoreductase KFYI subunit; Ndufc1
Gene Symbols NDUFC1
Host Species Rabbit
Immunogen Synthesized peptide derived from human NDUFC1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4717, 66377
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.