Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NDUFC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5103950
Description
Antibody detects endogenous levels of total NDUFC1.
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications
NDUFC1 | |
Polyclonal | |
Unconjugated | |
NDUFC1 | |
2310016K22Rik; CI-KFYI; complex I KFYI subunit; complex I-KFYI; KFYI; NADH dehydrogenase; NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 1; NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 1 (6kD, KFYI); NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 1, 6kDa; NADH dehydrogenase [ubiquinone] 1 subunit C1, mitochondrial; NADH:ubiquinone oxidoreductase subunit C1; NADH-ubiquinone oxidoreductase KFYI subunit; Ndufc1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
4717, 66377, 689938 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O43677, Q9CQY9 | |
NDUFC1 | |
A synthesized peptide derived from human NDUFC1(Accession O43677), corresponding to amino acid residues K43-E76. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction