Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30295520UL
Description
NDUFC2 Polyclonal specifically detects NDUFC2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockout Validated.Specifications
NDUFC2 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
4718 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, KnockDown | |
Unconjugated | |
B14.5b, CI-B14.5b, complex I subunit B14.5b, Complex I-B14.5b, HLC-1NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2 (14.5kD, B14.5b), Human lung cancer oncogene 1 protein, NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2, 14.5kDa, NADH dehydrogenase [ubiquinone] 1 subunit C2, NADHDH2, NADH-ubiquinone oxidoreductase subunit B14.5b | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human NDUFC2 (NP_004540.1). | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction