Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFS2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5112278

Description
NDUFS2 Polyclonal antibody specifically detects NDUFS2 in Cynomolgus Monkey, Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Specifications
NDUFS2 | |
Polyclonal | |
Unconjugated | |
Ndufs2 | |
AL033311; CI-49; CI-49kD; comp; complex 1, mitochondrial respiratory chain, 49-KD subunit; complex I 49kDa subunit; Complex I-49kD; EMBL:AAH82067.1}; NADH dehydrogenase (ubiquinone) Fe-S protein 2; NADH dehydrogenase (ubiquinone) Fe-S protein 2, 49kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 2, mitochondrial; NADH:ubiquinone oxidoreductase core subunit S2; NADH-coenzyme Q reductase; NADH-ubiquinone oxidoreductase 49 kDa subunit; NADH-ubiquinone oxidoreductase NDUFS2 subunit; Ndufs2; ndufs2 {ECO:0000312 | |
E. coli-derived Human NDUFS2 fragment | |
100 μL | |
Primary | |
Cynomolgus Monkey, Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 0.03% Proclin 300 | |
O75306 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
101925833, 4720 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction