Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFS2 Rabbit anti-Human, DyLight 488, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298489G
Description
NDUFS2 Polyclonal specifically detects NDUFS2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
NDUFS2 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 488 | |
CI-49, CI-49kD, complex 1, mitochondrial respiratory chain, 49-KD subunit, complex I 49kDa subunit, Complex I-49kD, EC 1.6.5.3, EC 1.6.99.3, EC 1.6.99.5, NADH dehydrogenase (ubiquinone) Fe-S protein 2 (49kD) (NADH-coenzyme Qreductase), NADH dehydrogenase (ubiquinone) Fe-S protein 2, 49kDa (NADH-coenzyme Qreductase), NADH dehydrogenase [ubiquinone] iron-sulfur protein 2, mitochondrial, NADH-ubiquinone oxidoreductase 49 kDa subunit, NADH-ubiquinone oxidoreductase NDUFS2 subunit | |
Produced in rabbits immunized with E. coli-derived Human NDUFS2 fragment. | |
0.1 mL | |
Core ESC Like Genes, Stem Cell Markers | |
4720 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction