Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFS3 Antibody (JU10-32), Novus Biologicals™
SDP

Rabbit Monoclonal Antibody

Supplier:  Novus Biologicals NBP275587

Catalog No. NBP275587


Only null left
Add to Cart

Description

Description

NDUFS3 Monoclonal specifically detects NDUFS3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications

Specifications

NDUFS3
Monoclonal
1 mg/mL
Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50
CI-30, CI-30kD, complex I 30kDa subunit, Complex I-30kD, EC 1.6.5.3, EC 1.6.99.3, EC 1.6.99.5, NADH dehydrogenase (ubiquinone) Fe-S protein 3 (30kD) (NADH-coenzyme Qreductase), NADH dehydrogenase (ubiquinone) Fe-S protein 3, 30kDa (NADH-coenzyme Qreductase), NADH dehydrogenase [ubiquinone] iron-sulfur protein 3, mitochondrial, NADH dehydrogenase-ubiquinone 30 kDa subunit, NADH-ubiquinone oxidoreductase 30 kDa subunit
Rabbit
25 kDa
100 μL
4722.0
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
JU10-32
Unconjugated
TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
NDUFS3
Synthetic peptide within Human NDUFS3 aa 215-264/ 264. (SwissProt: O75489 Human)
Protein A purified
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.