Learn More
Invitrogen™ NDUFS3 Monoclonal Antibody (17D95)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ 439200
Description
Positive control: Human heart mitochondria. In Flow Cytometry, this antibody gave a positive signal in methanol fixed/Tween permeabilised HepG2 cells.
This gene encodes one of the iron-sulfur protein components of mitochondrial NADH:ubiquinone oxidoreductase. Mutations in this gene are associated with Leigh syndrome resulting from mitochondrial complex I deficiency.
Specifications
| NDUFS3 | |
| Monoclonal | |
| 1 mg/mL | |
| HEPES buffered saline with 0.02% sodium azide | |
| O75489, P23709, Q9DCT2 | |
| NDUFS3 | |
| Bovine Heart Complex I. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Bovine | |
| Antibody | |
| IgG2a κ |
| Flow Cytometry, Western Blot | |
| 17D95 | |
| Unconjugated | |
| NDUFS3 | |
| 0610010M09Rik; CI-30; CI-30kD; complex I 30kDa subunit; complex I-30kD; hypothetical protein LOC550451; NADH dehydrogenase (ubiquinone); NADH dehydrogenase (ubiquinone) Fe-S protein 3; NADH dehydrogenase (ubiquinone) Fe-S protein 3, (NADH-coenzyme Q reductase); NADH dehydrogenase (ubiquinone) Fe-S protein 3, 30kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 3, mitochondrial; NADH dehydrogenase-ubiquinone 30 kDa subunit; NADH:ubiquinone oxidoreductase core subunit S3; NADH-ubiquinone oxidoreductase 30 kDa subunit; NDUFS3; zgc:112520 | |
| Mouse | |
| Antigen affinity chromatography | |
| RUO | |
| 287327, 295923, 4722, 68349 | |
| 4°C, do not freeze | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.