Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFS3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519118
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519118 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519118 Supplier Invitrogen™ Supplier No. PA519118
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.

This gene encodes one of the iron-sulfur protein components of mitochondrial NADH:ubiquinone oxidoreductase. Mutations in this gene are associated with Leigh syndrome resulting from mitochondrial complex I deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFS3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene NDUFS3
Gene Accession No. O75489
Gene Alias 0610010M09Rik; CI-30; CI-30kD; complex I 30kDa subunit; complex I-30kD; hypothetical protein LOC550451; NADH dehydrogenase (ubiquinone); NADH dehydrogenase (ubiquinone) Fe-S protein 3; NADH dehydrogenase (ubiquinone) Fe-S protein 3, (NADH-coenzyme Q reductase); NADH dehydrogenase (ubiquinone) Fe-S protein 3, 30kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 3, mitochondrial; NADH dehydrogenase-ubiquinone 30 kDa subunit; NADH:ubiquinone oxidoreductase core subunit S3; NADH-ubiquinone oxidoreductase 30 kDa subunit; NDUFS3; zgc:112520
Gene Symbols NDUFS3
Host Species Goat
Immunogen Synthetic peptide sequence (DTRPTVRPRNDVAHK) corresponding to the internal amino acids of NDUFS3 (aa 42-57).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4722
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.