Learn More
Invitrogen™ NDUFS3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA529747
Description
Recommended positive controls: 293T, THP-1, HL-60, Jurkat, NCI-H929. Predicted reactivity: Mouse (92%), Rat (91%), Zebrafish (84%), Xenopus laevis (85%), Pig (93%), Chicken (90%), Rhesus Monkey (98%), Chimpanzee (99%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes one of the iron-sulfur protein components of mitochondrial NADH:ubiquinone oxidoreductase. Mutations in this gene are associated with Leigh syndrome resulting from mitochondrial complex I deficiency.
Specifications
NDUFS3 | |
Polyclonal | |
Unconjugated | |
NDUFS3 | |
0610010M09Rik; CI-30; CI-30kD; complex I 30kDa subunit; complex I-30kD; hypothetical protein LOC550451; NADH dehydrogenase (ubiquinone); NADH dehydrogenase (ubiquinone) Fe-S protein 3; NADH dehydrogenase (ubiquinone) Fe-S protein 3, (NADH-coenzyme Q reductase); NADH dehydrogenase (ubiquinone) Fe-S protein 3, 30kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 3, mitochondrial; NADH dehydrogenase-ubiquinone 30 kDa subunit; NADH:ubiquinone oxidoreductase core subunit S3; NADH-ubiquinone oxidoreductase 30 kDa subunit; NDUFS3; zgc:112520 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4722 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.54 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O75489 | |
NDUFS3 | |
Recombinant protein encompassing a sequence within the center region of human NDUFS3. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.