Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ndufs4 Antibody (9H1), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | Ndufs4 |
---|---|
Clone | 9H1 |
Dilution | Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 |
Applications | Flow Cytometry, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
Ndufs4 Monoclonal antibody specifically detects Ndufs4 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunohistochemistrySpecifications
Ndufs4 | |
Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
AQDQ, AQDQmitochondrial, CI-18 kDa, NADH dehydrogenase (ubiquinone) Fe-S protein 4 (18kD) (NADH-coenzyme Qreductase), NADH dehydrogenase (ubiquinone) Fe-S protein 4, 18kDa (NADH-coenzyme Qreductase), NADH-coenzyme Q reductase, 18-KD, NADH-ubiquinone oxidoreductase 18 kDa subunit | |
A synthesized peptide derived from Human Ndufs4 [UniProt O43181] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
9H1 | |
Flow Cytometry, ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
4724 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title