Learn More
Invitrogen™ NDUFS4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592940
Description
Immunogen sequence: MAAVSMSVVL RQTLWRRRAV AVAALSVSRV PTRSLRTSTW RLAQDQTQDT QLITVDEKLD ITTLTGVPEE HIKTRKVRIF VPARNNMQSG VNNTKKWKME FDTRERWENP LMGWASTADP LSNMVLTFST KEDAVSFAEK NGWSYDIEER KVPKPKSKSY GANFSWNKRT RVSTK; Positive Samples: HepG2, 293T, mouse brain, mouse heart, SW620, MCF7, Mouse bone marrow, Mouse liver, Rat brain, Rat liver; Cellular Location: Matrix side, Mitochondrion inner membrane, Peripheral membrane protein.
This gene encodes an accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), or NADH:ubiquinone oxidoreductase, the first multi-subunit enzyme complex of the mitochondrial respiratory chain. Complex I plays a vital role in cellular ATP production, the primary source of energy for many crucial processes in living cells. It removes electrons from NADH and passes them by a series of different protein-coupled redox centers to the electron acceptor ubiquinone. In well-coupled mitochondria, the electron flux leads to ATP generation via the building of a proton gradient across the inner membrane. Complex I is composed of at least 41 subunits, of which 7 are encoded by the mitochondrial genome and the remainder by nuclear genes.
Specifications
NDUFS4 | |
Polyclonal | |
Unconjugated | |
NDUFS4 | |
6720411N02Rik; AQDQ; C1-18k; CI-18; CI-18 kDa; CI-AQDQ; complex I 18kDa subunit; complex I-18 kDa; Complex I-AQDQ; mitochondrial respiratory chain complex I (18-KD subunit); NADH dehydrogenase (ubiquinone) Fe-S protein 4; NADH dehydrogenase (ubiquinone) Fe-S protein 4, 18kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 4, mitochondrial; NADH:ubiquinone oxidoreductase subunit S4; NADH-ubiquinone oxidoreductase 18 kDa subunit; NDUFS4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
17993, 4724, 499529 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O43181, Q5XIF3, Q9CXZ1 | |
NDUFS4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-175 of human NDUFS4 (NP_0024861). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.