Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NDUFS4 Recombinant Rabbit Monoclonal Antibody (6T10F4)
GREENER_CHOICE

Catalog No. PIMA538178
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538178 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538178 Supplier Invitrogen™ Supplier No. MA538178
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: Mouse liver, Mouse heart, Rat brain, Rat kidney, Rat heart, HepG2 Immunogen sequence: MAAVSMSVVL RQTLWRRRAV AVAALSVSRV PTRSLRTSTW RLAQDQTQDT QLITVDEKLD ITTLTGVPEE HIKTRKVRIF VPARNNMQSG VNNTKKWKME.

This gene encodes an accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), or NADH:ubiquinone oxidoreductase, the first multi-subunit enzyme complex of the mitochondrial respiratory chain. Complex I plays a vital role in cellular ATP production, the primary source of energy for many crucial processes in living cells. It removes electrons from NADH and passes them by a series of different protein-coupled redox centers to the electron acceptor ubiquinone. In well-coupled mitochondria, the electron flux leads to ATP generation via the building of a proton gradient across the inner membrane. Complex I is composed of at least 41 subunits, of which 7 are encoded by the mitochondrial genome and the remainder by nuclear genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFS4
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 6T10F4
Concentration 1.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NDUFS4
Gene Accession No. O43181, Q5XIF3, Q9CXZ1
Gene Alias 6720411N02Rik; AQDQ; C1-18k; CI-18; CI-18 kDa; CI-AQDQ; complex I 18kDa subunit; complex I-18 kDa; Complex I-AQDQ; mitochondrial respiratory chain complex I (18-KD subunit); NADH dehydrogenase (ubiquinone) Fe-S protein 4; NADH dehydrogenase (ubiquinone) Fe-S protein 4, 18kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 4, mitochondrial; NADH:ubiquinone oxidoreductase subunit S4; NADH-ubiquinone oxidoreductase 18 kDa subunit; NDUFS4
Gene Symbols NDUFS4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human NDUFS4 (O43181).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17993, 4724, 499529
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.