Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDUFS5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577264
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577264 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577264 Supplier Thermo Scientific Supplier No. PA577264
Add to Cart
Add to Cart

NDUFS5 Polyclonal Antibody

NDUFS5 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFS5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene NDUFS5
Gene Accession No. O43920
Gene Alias AI256693; CI-15 kDa; CI15K; CI-15k; Complex I-15 kDa; NADH dehydrogenase (ubiquinone) Fe-S protein 5; NADH dehydrogenase (ubiquinone) Fe-S protein 5, 15kDa (NADH-coenzyme Q reductase); NADH dehydrogenase (ubiquinone) Fe-S protein 5b, 15kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 5; NADH:ubiquinone oxidoreductase 15 kDa IP subunit; NADH:ubiquinone oxidoreductase subunit S5; NADH-ubiquinone oxidoreductase 15 kDa subunit; Ndufs5; Ndufs5b
Gene Symbols NDUFS5
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 51-106 of Human NDUFS5.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4725
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.