Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NDUFS7 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519343
Description
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
This gene encodes a protein that is a subunit of one of the complexes that forms the mitochondrial respiratory chain. This protein is one of over 40 subunits found in complex I, the nicotinamide adenine dinucleotide (NADH):ubiquinone oxidoreductase. This complex functions in the transfer of electrons from NADH to the respiratory chain, and ubiquinone is believed to be the immediate electron acceptor for the enzyme. Mutations in this gene cause Leigh syndrome due to mitochondrial complex I deficiency, a severe neurological disorder that results in bilaterally symmetrical necrotic lesions in subcortical brain regions.
Specifications
NDUFS7 | |
Polyclonal | |
Unconjugated | |
Ndufs7 | |
1010001M04Rik; CI-20; CI-20KD; complex I 20kDa subunit; complex I, mitochondrial respiratory chain, 20-KD subunit; complex I-20kD; FLJ45860; FLJ46880; MGC120002; MY017; NADH dehydrogenase (ubiquinone) Fe-S protein 7; NADH dehydrogenase (ubiquinone) Fe-S protein 7, 20kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 7, mitochondrial; NADH:ubiquinone oxidoreductase core subunit S7; NADH:ubiquinone oxidoreductase PSST subunit; NADH-coenzyme Q reductase; NADH-ubiquinone oxidoreductase 20 kDa subunit; NADH-ubiquinone oxidoreductase Fe-S protein 7; NDUFS7; PSST; PSST subunit | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
374291 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O75251 | |
Ndufs7 | |
Synthetic peptide sequence (SRGEYVVAKLD) corresponding to the internal amino acids of NDUFS7 (aa 58-69). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction