Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFS7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | NDUFS7 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
NDUFS7 Polyclonal specifically detects NDUFS7 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
NDUFS7 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
374291 | |
IgG | |
Affinity purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
CI-20, CI-20kD, complex I, mitochondrial respiratory chain, 20-KD subunit, Complex I-20kD, EC 1.6.5.3, EC 1.6.99.3, EC 1.6.99.5, FLJ45860, FLJ46880, MGC120002, MY017, NADH dehydrogenase (ubiquinone) Fe-S protein 7 (20kD) (NADH-coenzyme Qreductase), NADH dehydrogenase (ubiquinone) Fe-S protein 7, 20kDa (NADH-coenzyme Qreductase), NADH dehydrogenase [ubiquinone] iron-sulfur protein 7, mitochondrial, NADH:ubiquinone oxidoreductase PSST subunit, NADH-coenzyme Q reductase, NADH-ubiquinone oxidoreductase 20 kDa subunit, PSST subunit, PSSTcomplex I 20kDa subunit | |
Recombinant fusion protein containing a sequence corresponding to amino acids 39-213 of human NDUFS7 (NP_077718.3). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title