Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFS7 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS11734R

Description
This gene encodes a protein that is a subunit of one of the complexes that forms the mitochondrial respiratory chain. This protein is one of over 40 subunits found in complex I, the nicotinamide adenine dinucleotide (NADH):ubiquinone oxidoreductase. This complex functions in the transfer of electrons from NADH to the respiratory chain, and ubiquinone is believed to be the immediate electron acceptor for the enzyme. Mutations in this gene cause Leigh syndrome due to mitochondrial complex I deficiency, a severe neurological disorder that results in bilaterally symmetrical necrotic lesions in subcortical brain regions.Specifications
NDUFS7 | |
Polyclonal | |
Unconjugated | |
Ndufs7 | |
1010001M04Rik; CI-20; CI-20kD; complex I 20kDa subunit; complex I, mitochondrial respiratory chain, 20-KD subunit; Complex I-20kD; FLJ45860; FLJ46880; MGC120002; MY017; NADH dehydrogenase (ubiquinone) Fe-S protein 7; NADH dehydrogenase (ubiquinone) Fe-S protein 7, 20kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 7, mitochondrial; NADH:ubiquinone oxidoreductase core subunit S7; NADH:ubiquinone oxidoreductase PSST subunit; NADH-coenzyme Q reductase; NADH-ubiquinone oxidoreductase 20 kDa subunit; NADH-ubiquinone oxidoreductase Fe-S protein 7; NDUFS7; PSST; PSST subunit | |
Rabbit | |
Protein A | |
RUO | |
374291 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
O75251 | |
Ndufs7 | |
KLH conjugated synthetic peptide derived from human NDUFS7. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction