Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDUFS8 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP149847
Description
NDUFS8 Polyclonal specifically detects NDUFS8 in Mouse, Rat samples. It is validated for Western Blot, Peptide ELISA.Specifications
NDUFS8 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
CI-23k, CI-23kD, CI23KD, complex I 23kDa subunit, Complex I-23kD, EC 1.6.5.3, EC 1.6.99.3, EC 1.6.99.5, NADH dehydrogenase (ubiquinone) Fe-S protein 8 (23kD) (NADH-coenzyme Qreductase), NADH dehydrogenase (ubiquinone) Fe-S protein 8, 23kDa (NADH-coenzyme Qreductase), NADH dehydrogenase [ubiquinone] iron-sulfur protein 8, mitochondrial, NADH-ubiquinone oxidoreductase 23 kDa subunit, TYKY, TYKY subunit | |
Goat | |
Affinity Purified | |
RUO | |
4728 | |
Mouse, Rat | |
IgG |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:4000 | |
NP_002487.1 | |
NDUFS8 | |
Peptide with sequence C-EAEIAANIQAD corresponding to internal region according to NP_002487.1. | |
0.1 mg | |
Primary | |
This antibody recognizes and epitope within the internal region.This antibody is specific to the internal region. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction