Learn More
Invitrogen™ NDUFS8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA522327
Description
Recommended positive controls: HepG2, mouse brain, rat brain. Predicted reactivity: Mouse (95%), Rat (96%), Xenopus laevis (95%), Dog (96%), Rhesus Monkey (96%), Chimpanzee (100%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Ndufs8 gene ontology annotations related to this gene include 4 iron, 4 sulfur cluster binding; oxidoreductase activity, acting on NAD(P)H.
Specifications
NDUFS8 | |
Polyclonal | |
Unconjugated | |
NDUFS8 | |
BC021616; CI-23k; CI23KD; CI-23kD; complex I 23kDa subunit; complex I-23kD; NADH dehydrogenase (ubiquinone) Fe-S protein 8; NADH dehydrogenase (ubiquinone) Fe-S protein 8, 23kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 8, mitochondrial; NADH:ubiquinone oxidoreductase core subunit S8; NADH-ubiquinone oxidoreductase 23 kDa subunit; NDUFS8; TYKY; TYKY subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
225887, 293652, 4728 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.35 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O00217, Q8K3J1 | |
NDUFS8 | |
Recombinant protein encompassing a sequence within the center region of human NDUFS8. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.